siRNA Details
Gene Symbol: ZNF557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044387.1 | NP_001037852.1 | 8 | 1472 | ||
NM_001044388.1 | NP_001037853.1 | 8 | 1463 | |||
NM_024341.2 | NP_077317.2 | 8 | 1484 | |||
GenBank mRNA | AK095524.1 | - | 8 | 1217 | ||
AK297283.1 | - | 8 | 1241 | |||
AK315405.1 | - | 8 | 1501 | |||
BC003020.1 | - | 8 | 1234 | 2822966 | ||
BC031208.1 | - | 8 | 1241 | |||
BC058040.1 | - | 8 | 1213 |
