siRNA Details
Gene Symbol: C3orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171747.1 | NP_001165218.1 | 1 | 162 | ||
NM_024616.2 | NP_078892.2 | 1 | 162 | |||
GenBank mRNA | AK026839.1 | - | 1 | 106 | ||
AK298076.1 | - | 1 | 60 | |||
AK303588.1 | - | 1 | 162 | |||
AY830714.1 | - | 1 | 108 | |||
BC017064.1 | - | 1 | 95 | 3885219 | ||
DB159226.1 | - | 1 | 162 |
