siRNA Details
Gene Symbol: ZNF329 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024620.3 | NP_078896.3 | 4 | 969 | ||
XM_006723381.2 | XP_006723444.1 | 3 | 906 | |||
XM_006723382.3 | XP_006723445.1 | 2 | 915 | |||
XM_006723383.3 | XP_006723446.1 | 2 | 1420 | |||
XM_006723384.3 | XP_006723447.1 | 2 | 1119 | |||
XM_011527310.2 | XP_011525612.1 | 5 | 1353 | |||
XM_011527311.2 | XP_011525613.1 | 4 | 1179 | |||
XM_011527312.2 | XP_011525614.1 | 4 | 944 | |||
XM_011527313.2 | XP_011525615.1 | 4 | 1256 | |||
XM_011527314.2 | XP_011525616.1 | 4 | 1235 | |||
XM_011527315.1 | XP_011525617.1 | 4 | 1315 | |||
XM_011527316.2 | XP_011525618.1 | 2 | 795 | |||
XM_017027307.1 | XP_016882796.1 | 4 | 1619 | |||
XM_017027308.1 | XP_016882797.1 | 3 | 1525 | |||
XM_017027309.1 | XP_016882798.1 | 3 | 1280 | |||
XM_017027310.1 | XP_016882799.1 | 3 | 1581 | |||
XM_017027311.1 | XP_016882800.1 | 2 | 1092 | |||
GenBank mRNA | AK022648.1 | - | 1 | 262 | ||
AK090893.1 | - | 4 | 1315 | |||
AK095349.1 | - | 5 | 1410 | |||
AK096542.1 | - | 2 | 776 | |||
BC047501.1 | - | 4 | 966 | 5266873 |
