siRNA Details
Gene Symbol: KIAA0319L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024874.4 | NP_079150.3 | 2, 3 | 388 | ||
XM_006710907.1 | XP_006710970.1 | 2, 3 | 187 | |||
XM_006710909.1 | XP_006710972.1 | 2, 3 | 279 | |||
XM_011542179.2 | XP_011540481.1 | 2, 3 | 519 | |||
XM_011542180.1 | XP_011540482.1 | 2, 3 | 357 | |||
XM_017002366.1 | XP_016857855.1 | 2, 3 | 357 | |||
XM_017002367.1 | XP_016857856.1 | 2, 3 | 519 | |||
XM_017002368.1 | XP_016857857.1 | 2, 3 | 187 | |||
XM_017002369.1 | XP_016857858.1 | 2, 3 | 279 | |||
XM_017002370.1 | XP_016857859.1 | 2, 3 | 357 | |||
XM_017002371.1 | XP_016857860.1 | 2, 3 | 357 | |||
XM_017002372.1 | XP_016857861.1 | 2, 3 | 279 | |||
XM_017002373.1 | XP_016857862.1 | 2, 3 | 357 | |||
GenBank mRNA | AF275679.2 | - | 2, 3 | 337 | ||
AK302428.1 | - | 2, 3 | 398 | |||
AK314605.1 | - | 2, 3 | 336 | |||
AK316252.1 | - | 2, 3 | 516 | |||
AY163234.1 | - | 2, 3 | 337 | |||
BC014530.1 | - | 2, 3 | 320 | 3843301 | ||
BX471865.1 | - | 2, 3 | 357 | |||
DC369564.1 | - | 2, 3 | 390 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001035526, NM_001035525, NM_133886, AK031342, BY308757, and others.
Gene Ontology Categories:
