siRNA Details
Gene Symbol: POF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307940.1 | NP_001294869.1 | 14 | 1637 | ||
NM_024921.3 | NP_079197.3 | 14 | 1637 | |||
XM_005262203.3 | XP_005262260.1 | 14 | 1550 | |||
GenBank mRNA | AF309774.1 | - | 14 | 1637 | ||
AK025039.1 | - | 10 | 1230 | |||
AK025080.1 | - | 6 | 602 | |||
AK026445.1 | - | 2 | 130 | |||
AK225193.1 | - | 6 | 602 | |||
AK290360.1 | - | 14 | 1586 | |||
AK314295.1 | - | 14 | 1556 | |||
BC017500.1 | - | 14 | 1611 | 4905554 |
