siRNA Details
Gene Symbol: POF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307940.1 | NP_001294869.1 | 4 | 557 | ||
NM_024921.3 | NP_079197.3 | 4 | 557 | |||
XM_005262203.3 | XP_005262260.1 | 4 | 515 | |||
GenBank mRNA | AF309774.1 | - | 4 | 557 | ||
AK025039.1 | - | 4 | 506 | |||
AK290360.1 | - | 4 | 506 | |||
AK314295.1 | - | 4 | 476 | |||
BC017500.1 | - | 4 | 531 | 4905554 |
