siRNA Details
Gene Symbol: C14orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001102366.2 | NP_001095836.1 | 13 | 2090 | ||
NM_001102367.2 | NP_001095837.1 | 12 | 1856 | |||
NM_001102368.2 | NP_001095838.1 | 11 | 1689 | |||
NM_001102369.2 | NP_001095839.1 | 13 | 1809 | |||
NM_001286470.1 | NP_001273399.1 | 14 | 2384 | |||
NM_001286471.1 | NP_001273400.1 | 11 | 1925 | |||
NM_001286472.1 | NP_001273401.1 | 13 | 1860 | |||
NM_024952.7 | NP_079228.4 | 11 | 1919 | |||
GenBank mRNA | AB210031.1 | - | 12 | 1590 | ||
AK022564.1 | - | 11 | 1490 | |||
AK023200.1 | - | 13 | 1607 | |||
AK024603.1 | - | 5 | 716 | |||
AK091394.1 | - | 12 | 1279 | |||
AK097294.1 | - | 11 | 1639 | |||
AK098752.1 | - | 11 | 1640 | |||
AK123532.1 | - | 14 | 2027 | |||
AK222531.1 | - | 5 | 715 | |||
AY358516.1 | - | 11 | 1925 | |||
AY927602.1 | - | LIT3593 AY927602 |
2 | 124 | ||
BC010614.2 | - | 12 | 1805 | 4157432 | ||
BC065558.1 | - | 13 | 1713 | 6138321 | ||
BX161393.1 | - | 11 | 1618 | |||
BX247989.1 | - | 13 | 1788 | |||
BX248294.1 | - | 9 | 1291 | |||
BX537970.1 | - | 11 | 1636 | |||
CR457289.1 | - | 9 | 1337 |
