siRNA Details
Gene Symbol: FAAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025161.5 | NP_079437.5 | 5 | 1651 | ||
NR_033338.1 | 5 | 1847 | ||||
XM_006722111.2 | XP_006722174.1 | 5 | 1646 | |||
XM_006722112.3 | XP_006722175.1 | 5 | 1797 | |||
XM_017025162.1 | XP_016880651.1 | 5 | 1672 | |||
XM_017025163.1 | XP_016880652.1 | 5 | 1568 | |||
GenBank mRNA | AK054713.1 | - | 1 | 995 | ||
AL834374.1 | - | 1 | 136 | |||
BC008883.2 | - | 5 | 1820 | |||
BC117139.1 | - | 3 | 1171 | 40125690 | ||
BC117141.1 | - | 3 | 1171 | 40125692 | ||
BC143861.1 | - | 3 | 1171 | |||
DQ989324.1 | - | 5 | 1633 |
