siRNA Details
Gene Symbol: ZNF430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172671.1 | NP_001166142.1 | 5 | 1714 | ||
NM_025189.3 | NP_079465.3 | 5 | 1717 | |||
GenBank mRNA | AK023721.1 | - | 5 | 1696 | ||
AK290475.1 | - | 5 | 1717 | |||
AL832422.1 | - | 1 | 602 | |||
AY269787.1 | - | 5 | 1646 | |||
BC111776.1 | - | 5 | 1602 | 40033720 |
