siRNA Details
Gene Symbol: ZNF451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257273.1 | NP_001244202.1 | 4 | 1908 | ||
XM_011514463.2 | XP_011512765.1 | 3 | 1665 | |||
GenBank mRNA | AB051489.1 | - | 1 | 1860 | ||
AK302577.1 | - | 4 | 1753 | |||
BC042450.1 | - | 2 | 1574 | |||
BC058853.1 | - | 2 | 1574 |
