siRNA Details
Gene Symbol: C1orf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030806.3 | NP_110433.1 | 2, 3 | 579 | ||
GenBank mRNA | AF312864.1 | - | 2, 3 | 501 | ||
AK056487.1 | - | 2, 3 | 315 | |||
AK312062.1 | - | 1, 2 | 216 | |||
AY258286.1 | - | 2, 3 | 492 | |||
BC028567.2 | - | 2, 3 | 537 | 3635521 | ||
BG823539.1 | - | 2, 3 | 569 |
