siRNA Details
Gene Symbol: TRAPPC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160372.3 | NP_001153844.1 | 16 | 2504 | ||
NM_001321646.1 | NP_001308575.1 | 15 | 2477 | |||
NM_031466.7 | NP_113654.4 | 16 | 2679 | |||
XM_011517326.2 | XP_011515628.1 | 15 | 2652 | |||
XM_011517328.2 | XP_011515630.1 | 16 | 2679 | |||
XM_011517330.2 | XP_011515632.1 | 8 | 1044 | |||
XM_017013893.1 | XP_016869382.1 | 16 | 2679 | |||
XM_017013894.1 | XP_016869383.1 | 9 | 1380 | |||
GenBank mRNA | AB067469.1 | - | 16 | 2454 | ||
AK026394.1 | - | 16 | 2496 | |||
AK027689.1 | - | 5 | 604 | |||
AK128755.1 | - | 16 | 2680 | |||
AK308815.1 | - | 5 | 775 | |||
AL833973.1 | - | 14 | 1779 | |||
AY190606.1 | - | 14 | 1758 | |||
BC003650.2 | - | 2 | 223 | |||
BC006206.2 | - | 14 | 1900 | 3538806 |
