siRNA Details
Gene Symbol: EMILIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032048.2 | NP_114437.2 | 4 | 2470 | ||
XM_017026038.1 | XP_016881527.1 | 4 | 2457 | |||
GenBank mRNA | AF270513.1 | - | 4 | 2470 | ||
AK090519.1 | - | 1 | 565 | |||
BC136541.1 | - | 4 | 2424 | 9020531 |
