siRNA Details
Gene Symbol: ZNF333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032433.2 | NP_115809.1 | 12 | 1433 | ||
XM_005260098.3 | XP_005260155.1 | 13 | 1746 | |||
XM_006722924.2 | XP_006722987.1 | 12 | 1510 | |||
XM_011528362.2 | XP_011526664.1 | 12 | 1474 | |||
XM_011528363.2 | XP_011526665.1 | 14 | 2574 | |||
XM_011528365.2 | XP_011526667.1 | 14 | 1915 | |||
XM_011528366.2 | XP_011526668.1 | 13 | 1743 | |||
XM_011528367.2 | XP_011526669.1 | 13 | 1756 | |||
XM_011528368.2 | XP_011526670.1 | 13 | 1663 | |||
XM_011528369.2 | XP_011526671.1 | 13 | 1676 | |||
XM_011528371.2 | XP_011526673.1 | 6 | 1840 | |||
XM_011528372.2 | XP_011526674.1 | 5 | 769 | |||
XM_017027367.1 | XP_016882856.1 | 12 | 1450 | |||
XM_017027368.1 | XP_016882857.1 | 11 | 1318 | |||
XM_017027369.1 | XP_016882858.1 | 13 | 3071 | |||
XM_017027370.1 | XP_016882859.1 | 13 | 3084 | |||
XM_017027371.1 | XP_016882860.1 | 13 | 1696 | |||
GenBank mRNA | AB058709.1 | - | 5 | 747 | ||
AF372702.1 | - | 12 | 1390 | |||
AK094028.1 | - | 13 | 1676 | |||
AK095142.1 | - | 1 | 315 | |||
AK122817.1 | - | 5 | 1314 | |||
AL832544.1 | - | 6 | 925 | |||
BX647475.1 | - | 12 | 1394 |
