siRNA Details
Gene Symbol: SMARCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003601.3 | NP_003592.3 | 17 | 2716 | ||
GenBank mRNA | AB010882.1 | - | 17 | 2456 | ||
AK302841.1 | - | 18 | 2351 | |||
AK316444.1 | - | 18 | 2518 | |||
BC023144.1 | - | 17 | 2467 | 4340921 |
