siRNA Details
Gene Symbol: FIBCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145106.1 | NP_001138578.1 | 6, 7 | 1051 | ||
NM_032843.4 | NP_116232.3 | 5, 6 | 1189 | |||
GenBank mRNA | AK292489.1 | - | 5, 6 | 958 | ||
AY358311.1 | - | 5, 6 | 946 | |||
BC032953.2 | - | 5, 6 | 1187 | 4811679 | ||
BC112892.1 | - | 5, 6 | 939 |
