siRNA Details
Gene Symbol: CCDC183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039374.4 | NP_001034463.4 | 10, 11 | 1157 | ||
GenBank mRNA | BC064582.1 | - | 10, 11 | 1143 | ||
BC137317.1 | - | 10, 11 | 1132 | 9021314 | ||
BC137318.1 | - | 10, 11 | 1132 | 9021315 |
