siRNA Details
Gene Symbol: GAL3ST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033036.2 | NP_149025.1 | 3 | 1362 | ||
XM_017018519.1 | XP_016874008.1 | 2 | 1276 | |||
GenBank mRNA | AB053232.1 | - | 3 | 1362 | ||
AY026481.1 | - | 2 | 1240 | |||
BC113552.1 | - | 2 | 1148 | 8322604 | ||
BC113554.1 | - | 2 | 1148 | 8322606 |
