siRNA Details
Gene Symbol: ZNF804A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194250.1 | NP_919226.1 | 4 | 3865 | ||
GenBank mRNA | AF052145.1 | - | 1 | 597 | ||
AK055275.1 | - | 1 | 1598 | |||
AK127762.1 | - | 3 | 3042 | |||
AK295280.1 | - | 1 | 1329 | |||
AY327407.1 | - | 4 | 3865 | |||
BC136602.1 | - | 4 | 3523 | 9020592 | ||
BC150198.1 | - | 4 | 3722 | 40148778 |
