siRNA Details
Gene Symbol: TMEM129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127266.1 | NP_001120738.1 | 2 | 1016 | ||
NM_138385.3 | NP_612394.1 | 2 | 1016 | |||
XM_005248038.4 | XP_005248095.1 | 2 | 888 | |||
XM_005248039.4 | XP_005248096.1 | 2 | 457 | |||
XM_011513594.2 | XP_011511896.1 | 2 | 466 | |||
XM_017008834.1 | XP_016864323.1 | 2 | 753 | |||
GenBank mRNA | AK303880.1 | - | 2 | 329 | ||
BC009331.2 | - | 2 | 1016 | |||
BC018921.2 | - | 2 | 1016 | |||
BC126370.1 | - | 3 | 906 | 8992086 |
