siRNA Details
Gene Symbol: C1orf105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300760.1 | NP_001287689.1 | 2 | 310 | ||
NM_139240.3 | NP_640333.3 | 4 | 409 | |||
XM_005245608.2 | XP_005245665.1 | 4 | 273 | |||
XM_005245610.3 | XP_005245667.1 | 3 | 190 | |||
XM_006711630.2 | XP_006711693.1 | 2 | 96 | |||
XM_011510152.2 | XP_011508454.1 | 6 | 2618 | |||
XM_011510153.2 | XP_011508455.1 | 5 | 2581 | |||
XM_011510154.2 | XP_011508456.1 | 3 | 2451 | |||
XM_011510155.2 | XP_011508457.1 | 6 | 2618 | |||
XM_011510156.2 | XP_011508458.1 | 2 | 2360 | |||
XM_017002808.1 | XP_016858297.1 | 4 | 207 | |||
GenBank mRNA | AL035295.1 | - | 4 | 409 | ||
BC038410.1 | - | 4 | 407 | 5169137 |
