siRNA Details
Gene Symbol: BBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195304.1 | NP_001182233.1 | 5 | 690 | ||
NM_001195305.1 | NP_001182234.1 | 4 | 533 | |||
NM_001195306.1 | NP_001182235.1 | 4 | 379 | |||
NM_001195307.1 | NP_001182236.1 | 3 | 458 | |||
NM_001243783.1 | NP_001230712.1 | 3 | 440 | |||
GenBank mRNA | AK025724.1 | - | 4 | 353 | ||
AK125829.1 | - | 1 | 4537 | |||
AK307126.1 | - | 4 | 379 | |||
AK307314.1 | - | 5 | 566 | |||
BC104248.1 | - | 4 | 340 | |||
BC104249.1 | - | 4 | 342 | |||
BC105804.1 | - | 5 | 529 | |||
BC113000.1 | - | 4 | 342 | |||
BC113001.1 | - | 4 | 342 | |||
BI552279.1 | - | 3 | 272 |
