siRNA Details
Gene Symbol: ZNF516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014643.3 | NP_055458.1 | 6, 7 | 3753 | ||
XM_011526269.1 | XP_011524571.1 | 5, 6 | 3691 | |||
XM_011526270.2 | XP_011524572.1 | 5, 6 | 3954 | |||
XM_011526271.2 | XP_011524573.1 | 6, 7 | 4068 | |||
XM_011526272.2 | XP_011524574.1 | 6, 7 | 3789 | |||
XM_011526273.2 | XP_011524575.1 | 5, 6 | 3704 | |||
XM_011526274.2 | XP_011524576.1 | 5, 6 | 3710 | |||
XM_011526275.2 | XP_011524577.1 | 5, 6 | 4567 | |||
XM_017026097.1 | XP_016881586.1 | 6, 7 | 3824 | |||
GenBank mRNA | D86975.1 | - | 6, 7 | 3753 |
