siRNA Details
Gene Symbol: TECPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172631.2 | NP_001166102.1 | 5 | 802 | ||
NM_014844.4 | NP_055659.2 | 5 | 802 | |||
GenBank mRNA | AB002295.1 | - | 2 | 105 | ||
AB002327.1 | - | 5 | 751 | |||
AK127721.1 | - | 5 | 1235 | |||
AK296395.1 | - | 5 | 653 | |||
BC136647.1 | - | 4 | 599 | 9020637 | ||
BC142667.1 | - | 5 | 635 | 40148157 | ||
BC142715.1 | - | 5 | 635 | 40148843 | ||
BC151230.1 | - | 5 | 775 | 8860116 |
