siRNA Details
Gene Symbol: Camta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081557.3 | NP_001075026.1 | 13, 14 | 3376 | ||
GenBank mRNA | AK122383.1 | - | 8, 9 | 2802 | ||
AK132560.1 | - | 1, 2 | 181 | |||
AK147482.1 | - | 3, 4 | 522 | |||
BC150740.1 | - | 12, 13 | 3225 | 9087651 |

More Information
Other Species Targeted

Gene Ontology Categories:
