siRNA Details
Gene Symbol: Camta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081557.3 | NP_001075026.1 | 16 | 4192 | ||
GenBank mRNA | AK122383.1 | - | 11 | 3618 | ||
AK132560.1 | - | 4 | 996 | |||
AK147482.1 | - | 6 | 999 | |||
AK168378.1 | - | 6 | 925 | |||
BC150740.1 | - | 15 | 3702 | 9087651 |

More Information
Other Species Targeted

Gene Ontology Categories:
