siRNA Details
Gene Symbol: Camta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081557.3 | NP_001075026.1 | 11, 12 | 3009 | ||
GenBank mRNA | AK122383.1 | - | 6, 7 | 2435 | ||
BC150740.1 | - | 10, 11 | 2858 | 9087651 |
