siRNA Details
Gene Symbol: Lrrc8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133897.2 | NP_598658.1 | 4 | 1721 | ||
GenBank mRNA | AB081508.1 | - | 4 | 1721 | ||
AK089024.1 | - | 4 | 1789 | |||
AK142337.1 | - | 4 | 1751 | |||
AK165201.1 | - | 1 | 741 | |||
AK169492.1 | - | 4 | 1784 | |||
AK171313.1 | - | 4 | 1803 | |||
BC025473.1 | - | 1 | 1242 | 5353064 | ||
BC026572.1 | - | 4 | 1731 | 4988595 |
