siRNA Details
Gene Symbol: Snx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028874.2 | NP_083150.1 | 10, 11 | 3098 | ||
GenBank mRNA | AK014388.1 | - | 4, 5 | 545 | ||
AK032107.1 | - | 6, 7 | 736 | |||
AK044307.1 | - | 9, 10 | 2142 | |||
AK122231.1 | - | 10, 11 | 3319 | |||
BC004635.1 | - | 10, 11 | 1570 | |||
BC051164.1 | - | 10, 11 | 3318 | |||
BC063262.1 | - | 10, 11 | 3098 | 6405897 |
