siRNA Details
Gene Symbol: A630052K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042023.1 | - | 1 | 752 |
Gene Symbol: Pan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252326.1 | NP_001239255.1 | 6 | 1145 | ||
NM_001252327.1 | NP_001239256.1 | 6 | 1091 | |||
NM_133992.3 | NP_598753.1 | 6 | 1145 | |||
GenBank mRNA | AK004773.2 | - | 6 | 1013 | ||
AK047887.1 | - | 6 | 1105 | |||
AK089001.1 | - | 6 | 1146 | |||
AK129196.1 | - | 6 | 1126 | |||
AK157800.1 | - | 6 | 1049 | |||
BC008521.1 | - | 6 | 1082 | |||
BC075686.1 | - | 6 | 1122 | 30615204 | ||
BC079841.1 | - | 6 | 1106 | 6417806 | ||
BG086688.2 | - | 3 | 376 |
