siRNA Details
Gene Symbol: Pwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133993.3 | NP_598754.2 | 11, 12 | 1197 | ||
GenBank mRNA | AK003983.1 | - | 1, 2 | 101 | ||
AK009972.1 | - | 11, 12 | 1164 | |||
AK012649.1 | - | 5, 6 | 440 | |||
AK082158.1 | - | 11, 12 | 1139 | |||
AK149699.1 | - | 11, 12 | 1142 | |||
BC023137.1 | - | 11, 12 | 1147 | 3986908 |
