siRNA Details
Gene Symbol: Cep57l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243074.1 | NP_001230003.1 | 5 | 586 | ||
NM_001243075.1 | NP_001230004.1 | 3 | 277 | |||
NM_026643.5 | NP_080919.3 | 5 | 504 | |||
NM_029132.2 | NP_083408.1 | 5 | 572 | |||
GenBank mRNA | AK010530.1 | - | 5 | 523 | ||
AK012710.1 | - | 3 | 223 | |||
AK015619.1 | - | 4 | 487 | |||
BC021375.1 | - | 5 | 596 | 5038957 | ||
BC138230.1 | - | 5 | 507 | 8861249 | ||
BQ553570.1 | - | 4 | 461 | |||
CD354087.1 | - | 2 | 254 |

More Information
Other Species Targeted

Gene Ontology Categories:
