siRNA Details
Gene Symbol: Cep57l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243074.1 | NP_001230003.1 | 6, 7 | 739 | ||
NM_001243075.1 | NP_001230004.1 | 4, 5 | 430 | |||
NM_026643.5 | NP_080919.3 | 6, 7 | 657 | |||
NM_029132.2 | NP_083408.1 | 6, 7 | 725 | |||
GenBank mRNA | AF397908.1 | - | 6, 7 | 653 | ||
AK010530.1 | - | 6, 7 | 676 | |||
AK012710.1 | - | 4, 5 | 376 | |||
AK015619.1 | - | 5, 6 | 640 | |||
BC021375.1 | - | 6, 7 | 749 | 5038957 | ||
BC138230.1 | - | 6, 7 | 660 | 8861249 | ||
CD354087.1 | - | 3, 4 | 407 |
