siRNA Details
Gene Symbol: Gldc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138595.2 | NP_613061.1 | 21 | 2739 | ||
GenBank mRNA | AK010327.1 | - | 1 | 50 | ||
AK080789.1 | - | 21 | 2711 | |||
AK080937.1 | - | 21 | 2729 | |||
BC017135.1 | - | 21 | 2716 | 4511573 |
