siRNA Details
Gene Symbol: Kdm3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038695.3 | NP_001033784.2 | 20 | 3204 | ||
NM_173001.3 | NP_766589.1 | 19 | 3231 | |||
GenBank mRNA | AK129204.1 | - | 20 | 3192 | ||
AK166797.1 | - | 10 | 1627 | |||
BC026605.1 | - | 5 | 819 | |||
BC031158.1 | - | 8 | 1291 | |||
BC031200.1 | - | 8 | 1275 | |||
BC031909.1 | - | 3 | 359 | |||
BC032256.1 | - | 20 | 3180 | |||
BC049876.1 | - | 13 | 2274 | |||
BC059264.1 | - | 19 | 3114 | 6409698 | ||
DQ323991.1 | - | 16 | 2669 | |||
KJ461739.1 | - | 10 | 1535 |
