siRNA Details
Gene Symbol: Arrdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042591.1 | NP_001036056.1 | 5, 6 | 1089 | ||
GenBank mRNA | AK084951.1 | - | 5, 6 | 1100 | ||
AK088288.1 | - | 5, 6 | 1104 | |||
AK122499.1 | - | 5, 6 | 1076 | |||
AK163364.1 | - | 5, 6 | 634 | |||
BC023838.1 | - | 2, 3 | 307 | |||
BC031125.1 | - | 3, 4 | 467 | |||
BC044762.1 | - | 5, 6 | 1087 | |||
BC054844.1 | - | 5, 6 | 1089 | 6493441 | ||
BC098474.1 | - | 5, 6 | 1083 | 30094818 | ||
DQ861994.1 | - | 5, 6 | 834 |
