siRNA Details
Gene Symbol: Catsperd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175350.3 | NP_780559.2 | 20 | 1939 | ||
GenBank mRNA | AK077082.1 | - | 11 | 1000 | ||
BC115864.1 | - | 9 | 892 | |||
BC116633.1 | - | 10 | 1006 | 40056655 | ||
HQ441159.1 | - | 20 | 1931 | |||
HQ441160.1 | - | 17 | 1827 |
