siRNA Details
Gene Symbol: Rapgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039086.1 | NP_001034175.1 | 25 | 3714 | ||
NM_001039087.1 | NP_001034176.1 | 25 | 3732 | |||
NM_054050.2 | NP_473391.1 | 23 | 3318 | |||
GenBank mRNA | AF348669.1 | - | 23 | 3274 | ||
AK077034.1 | - | 15 | 1704 | |||
AK147468.1 | - | 25 | 3733 | |||
AK147470.1 | - | 25 | 3733 | |||
AK147506.1 | - | 25 | 3701 | |||
AK147696.1 | - | 25 | 3694 | |||
BC034081.1 | - | 6 | 483 | |||
BC039756.1 | - | 11 | 1045 | |||
BC129881.1 | - | 24 | 3425 | 40110588 |
