siRNA Details
Gene Symbol: Rapgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039086.1 | NP_001034175.1 | 18 | 2972 | ||
NM_001039087.1 | NP_001034176.1 | 18 | 2990 | |||
NM_054050.2 | NP_473391.1 | 16 | 2576 | |||
GenBank mRNA | AF348669.1 | - | 16 | 2532 | ||
AK077034.1 | - | 8 | 962 | |||
AK147468.1 | - | 18 | 2991 | |||
AK147470.1 | - | 18 | 2991 | |||
AK147506.1 | - | 18 | 2959 | |||
AK147696.1 | - | 18 | 2952 | |||
BC039756.1 | - | 4 | 303 | |||
BC129881.1 | - | 17 | 2683 | 40110588 |
