siRNA Details
Gene Symbol: Uhrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144873.2 | NP_659122.2 | 7 | 1491 | ||
GenBank mRNA | AB116653.1 | - | 7 | 1226 | ||
AF274047.1 | - | 7 | 1401 | |||
AK080925.1 | - | 7 | 1532 | |||
BC060241.1 | - | 7 | 1491 | 6334856 |
