siRNA Details
Gene Symbol: Chd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177224.2 | NP_796198.1 | 34 | 7243 | ||
GenBank mRNA | AB093226.3 | - | 21 | 3935 | ||
BC052896.1 | - | 3 | 274 | |||
BC145219.1 | - | 32 | 6939 | 9053770 | ||
BC145220.1 | - | 31 | 6736 | |||
DQ127229.2 | - | 34 | 7243 |
