siRNA Details
Gene Symbol: F11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF356627.1 | - | 12 | 1492 | ||
AK005546.1 | - | 12 | 1504 | |||
BC019485.1 | - | 12 | 1481 | 4194672 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF356627.1 | - | 12 | 1492 | ||
AK005546.1 | - | 12 | 1504 | |||
BC019485.1 | - | 12 | 1481 | 4194672 |