siRNA Details
Gene Symbol: Dnah5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133365.3 | NP_579943.3 | 14 | 2189 | ||
GenBank mRNA | AF466704.1 | - | 14 | 2190 | ||
AK054152.1 | - | 14 | 2191 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnah5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133365.3 | NP_579943.3 | 14 | 2189 | ||
GenBank mRNA | AF466704.1 | - | 14 | 2190 | ||
AK054152.1 | - | 14 | 2191 |