siRNA Details
Gene Symbol: Prdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081355.3 | NP_001074824.3 | 8 | 4028 | ||
NM_001256380.1 | NP_001243309.1 | 8 | 4028 | |||
GenBank mRNA | BC043456.1 | - | 1 | 500 | ||
BC052515.1 | - | 1 | 231 | |||
BC151062.1 | - | 7 | 3941 | 9087975 |
