siRNA Details
Gene Symbol: Prdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081355.3 | NP_001074824.3 | 8 | 2873 | ||
NM_001256380.1 | NP_001243309.1 | 8 | 2873 | |||
GenBank mRNA | BC151062.1 | - | 7 | 2786 | 9087975 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001077648, XM_008764255, XM_006239335, XM_006239337, U17837.
Gene Ontology Categories:
