siRNA Details
Gene Symbol: Cldn16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053241.5 | NP_444471.1 | 2 | 539 | ||
GenBank mRNA | AB009016.1 | - | 2 | 201 | ||
AF323748.1 | - | 2 | 329 | |||
AK085268.1 | - | 2 | 540 | |||
AK085333.1 | - | 2 | 515 | |||
BC115574.2 | - | 3 | 318 | 40102178 | ||
BC115575.2 | - | 3 | 318 | 40102179 |
