siRNA Details
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053261.2 | NP_444491.1 | 3, 4 | 512 | ||
GenBank mRNA | AF353730.1 | - | 3, 4 | 511 | ||
AK154489.1 | - | 3, 4 | 550 | |||
AK154845.1 | - | 3, 4 | 483 | |||
AK170962.1 | - | 3, 4 | 550 | |||
AK171626.1 | - | 3, 4 | 485 |
