siRNA Details
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053261.2 | NP_444491.1 | 7 | 917 | ||
GenBank mRNA | AF353730.1 | - | 7 | 916 | ||
AK154489.1 | - | 7 | 955 | |||
AK154845.1 | - | 7 | 888 | |||
AK170962.1 | - | 7 | 955 | |||
AK171626.1 | - | 7 | 890 | |||
BC011093.1 | - | 7 | 892 | 4012163 |
