siRNA Details
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053261.2 | NP_444491.1 | 8 | 1016 | ||
GenBank mRNA | AF353730.1 | - | 8 | 1015 | ||
AK154489.1 | - | 8 | 1054 | |||
AK154845.1 | - | 8 | 987 | |||
AK170962.1 | - | 8 | 1054 | |||
AK171626.1 | - | 8 | 989 | |||
BC011093.1 | - | 8 | 991 | 4012163 |
