siRNA Details
Gene Symbol: Mta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171052.1 | NP_001164523.1 | 13 | 1311 | ||
NM_001171053.1 | NP_001164524.1 | 13 | 1314 | |||
NM_001171054.1 | NP_001164525.1 | 13 | 1311 | |||
NM_054082.2 | NP_473423.1 | 13 | 1308 | |||
GenBank mRNA | AF288138.1 | - | 13 | 1262 | ||
AK003306.1 | - | 791 | ||||
AK129318.1 | - | 13 | 1243 | |||
AK145942.1 | - | 13 | 1256 | |||
AK146721.1 | - | 13 | 1264 | |||
AK146905.1 | - | 13 | 1264 | |||
AK147197.1 | - | 13 | 1254 | |||
AK154856.1 | - | 13 | 1251 | |||
AK158866.1 | - | 12 | 1174 | |||
BC022124.1 | - | 13 | 1275 | 5097283 | ||
BC099960.1 | - | 13 | 1231 | 30023887 | ||
CA751287.1 | - | 7 | 654 |
